Assay Details
| Gene Symbol: PLAUR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005376.2 | NP_001005376.1 | 3 - 4 | 534 | 85 | |
| NM_001005377.2 | NP_001005377.1 | 3 - 4 | 534 | 85 | ||
| NM_001301037.1 | NP_001287966.1 | 3 - 4 | 534 | 85 | ||
| NM_002659.3 | NP_002650.1 | 3 - 4 | 534 | 85 | ||
| XM_005258990.4 | XP_005259047.1 | 3 - 4 | 356 | 85 | ||
| XM_011527028.2 | XP_011525330.1 | 3 - 4 | 356 | 85 | ||
| XM_011527029.1 | XP_011525331.1 | 3 - 4 | 356 | 85 | ||
| XM_011527030.2 | XP_011525332.1 | 3 - 4 | 356 | 85 | ||
| XM_011527031.2 | XP_011525333.1 | 3 - 4 | 356 | 85 | ||
| XM_017026872.1 | XP_016882361.1 | 3 - 4 | 769 | 85 | ||
| GenBank mRNA | A18757.1 | - | 3 - 4 | 354 | 85 | |
| AF257789.1 | - | 3 - 4 | 308 | 85 | ||
| AK290774.1 | - | 3 - 4 | 354 | 85 | ||
| AY029180.1 | - | 3 - 4 | 354 | 85 | ||
| BC002788.2 | - | 3 - 4 | 373 | 3617894 | 85 | |
| CA495321.1 | - | 3 - 4 | 534 | 85 | ||
| CF126152.1 | - | 3 - 4 | 341 | 85 | ||
| CR456952.1 | - | 3 - 4 | 308 | 85 | ||
| M83246.1 | - | 3 - 4 | 325 | 85 | ||
| U08839.1 | - | 3 - 4 | 734 | 85 | ||
| X51675.1 | - | 3 - 4 | 354 | 85 | ||
| X74039.1 | - | 3 - 4 | 354 | 85 | ||
More Information
Set Membership: |
|

PLAUR
Genomic Map