Assay Details
| Gene Symbol: SHF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301170.1 | NP_001288099.1 | 3 - 4 | 868 | 101 | |
| GenBank mRNA | BX446025.2 | - | 3 - 4 | 553 | 101 | |
More Information
Set Membership: |
|

SHF
Genomic Map