Assay Details
| Gene Symbol: SS18L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301778.1 | NP_001288707.1 | 8 - 9 | 1029 | 55 | |
| NM_198935.2 | NP_945173.1 | 7 - 8 | 924 | 55 | ||
| NR_125980.1 | 8 - 9 | 1303 | 55 | |||
| NR_125981.1 | 7 - 8 | 924 | 55 | |||
| XM_005260389.3 | XP_005260446.1 | 7 - 8 | 879 | 55 | ||
| XM_005260391.1 | XP_005260448.1 | 8 - 9 | 1063 | 55 | ||
| XM_011528764.2 | XP_011527066.1 | 8 - 9 | 2055 | 55 | ||
| XM_011528767.1 | XP_011527069.1 | 6 - 7 | 865 | 55 | ||
| XM_017027784.1 | XP_016883273.1 | 7 - 8 | 1245 | 55 | ||
| GenBank mRNA | AB014593.1 | - | 7 - 8 | 849 | 55 | |
| AK125656.1 | - | 8 - 9 | 996 | 55 | ||
| AK291485.1 | - | 7 - 8 | 899 | 55 | ||
| AK299881.1 | - | 7 - 8 | 899 | 55 | ||
| AY203931.1 | - | 4 - 5 | 611 | 55 | ||
| BC034494.2 | - | 7 - 8 | 878 | 4823298 | 55 | |
| BC068993.1 | - | 8 - 9 | 1241 | 5094932 | 55 | |
| BX640848.1 | - | 8 - 9 | 1061 | 55 | ||
More Information
Set Membership: |
|

SS18L1
Genomic Map