Assay Details
| Gene Symbol: MAEA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017405.2 | NP_001017405.1 | 2 - 3 | 318 | 105 | |
| NM_001297431.1 | NP_001284360.1 | 2 - 3 | 318 | 105 | ||
| NM_001297432.1 | NP_001284361.1 | 2 - 3 | 456 | 105 | ||
| NM_001297433.1 | NP_001284362.1 | 2 - 3 | 392 | 105 | ||
| NM_005882.4 | NP_005873.2 | 2 - 3 | 318 | 105 | ||
| NR_123716.1 | 2 - 3 | 318 | 105 | |||
| XM_006713849.3 | XP_006713912.1 | 2 - 3 | 278 | 105 | ||
| XM_006713850.2 | XP_006713913.1 | 2 - 3 | 451 | 105 | ||
| XM_011513376.1 | XP_011511678.1 | 3 - 4 | 438 | 105 | ||
| XM_017007636.1 | XP_016863125.1 | 3 - 4 | 438 | 105 | ||
| GenBank mRNA | AK001088.1 | - | 2 - 3 | 278 | 105 | |
| AK022515.1 | - | 2 - 3 | 287 | 105 | ||
| AK022586.1 | - | 2 - 3 | 278 | 105 | ||
| AK298944.1 | - | 2 - 3 | 372 | 105 | ||
| AK301155.1 | - | 2 - 3 | 286 | 105 | ||
| AK308901.1 | - | 2 - 3 | 262 | 105 | ||
| AY236486.1 | - | 2 - 3 | 263 | 105 | ||
| BC001225.1 | - | 2 - 3 | 285 | 3533990 | 105 | |
| BC006470.1 | - | 2 - 3 | 241 | 105 | ||
| BT006957.1 | - | 2 - 3 | 222 | 105 | ||
| DC299400.1 | - | 2 - 3 | 395 | 105 | ||
| DC331820.1 | - | 2 - 3 | 319 | 105 | ||
| DC400017.1 | - | 2 - 3 | 456 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

MAEA
Genomic Map