Assay Details
| Gene Symbol: MAEA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017405.2 | NP_001017405.1 | 5 - 6 | 715 | 64 | |
| NM_001297430.1 | NP_001284359.1 | 4 - 5 | 511 | 64 | ||
| NM_001297432.1 | NP_001284361.1 | 5 - 6 | 853 | 64 | ||
| NM_001297433.1 | NP_001284362.1 | 5 - 6 | 789 | 64 | ||
| NM_005882.4 | NP_005873.2 | 4 - 5 | 592 | 64 | ||
| XM_006713849.3 | XP_006713912.1 | 5 - 6 | 675 | 64 | ||
| XM_006713850.2 | XP_006713913.1 | 5 - 6 | 848 | 64 | ||
| XM_011513376.1 | XP_011511678.1 | 6 - 7 | 835 | 64 | ||
| XM_011513377.1 | XP_011511679.1 | 5 - 6 | 753 | 64 | ||
| XM_017007636.1 | XP_016863125.1 | 6 - 7 | 835 | 64 | ||
| XM_017007637.1 | XP_016863126.1 | 5 - 6 | 752 | 64 | ||
| GenBank mRNA | AF084928.1 | - | 5 - 6 | 540 | 64 | |
| AK001088.1 | - | 5 - 6 | 675 | 64 | ||
| AK022586.1 | - | 4 - 5 | 552 | 64 | ||
| AK296174.1 | - | 4 - 5 | 468 | 64 | ||
| AK298944.1 | - | 5 - 6 | 769 | 64 | ||
| AK300572.1 | - | 6 - 7 | 612 | 64 | ||
| AK301155.1 | - | 5 - 6 | 683 | 64 | ||
| AY189687.1 | - | 5 - 6 | 628 | 64 | ||
| BC001225.1 | - | 5 - 6 | 682 | 3533990 | 64 | |
| BC006470.1 | - | 5 - 6 | 638 | 64 | ||
| BT006957.1 | - | 5 - 6 | 619 | 64 | ||
More Information
Set Membership: |
|

MAEA
Genomic Map