Assay Details
| Gene Symbol: CCL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006664.3 | NP_006655.1 | 1 - 2 | 128 | 90 | |
| GenBank mRNA | AB010445.1 | - | 1 - 2 | 128 | 90 | |
| AF082393.1 | - | 1 - 2 | 69 | 90 | ||
| AJ243542.1 | - | 1 - 2 | 93 | 90 | ||
| BC148263.1 | - | 1 - 2 | 91 | 8860203 | 90 | |
More Information
Set Membership: |
|

CCL27
Genomic Map