Assay Details
| Gene Symbol: PRPF40B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031698.2 | NP_001026868.2 | 4 - 5 | 475 | 76 | |
| NM_012272.2 | NP_036404.1 | 3 - 4 | 765 | 76 | ||
| XM_011538139.2 | XP_011536441.1 | 4 - 5 | 469 | 76 | ||
| XM_011538140.2 | XP_011536442.1 | 4 - 5 | 469 | 76 | ||
| XM_011538141.1 | XP_011536443.1 | 3 - 4 | 295 | 76 | ||
| XM_011538143.2 | XP_011536445.1 | 3 - 4 | 311 | 76 | ||
| XM_011538144.2 | XP_011536446.1 | 3 - 4 | 295 | 76 | ||
| XM_017019135.1 | XP_016874624.1 | 3 - 4 | 232 | 76 | ||
| XM_017019136.1 | XP_016874625.1 | 3 - 4 | 295 | 76 | ||
| XM_017019137.1 | XP_016874626.1 | 3 - 4 | 295 | 76 | ||
| GenBank mRNA | AF049525.1 | - | 2 - 3 | 129 | 76 | |
| AL834216.1 | - | 3 - 4 | 765 | 76 | ||
| BC031917.1 | - | 1 - 2 | 71 | 76 | ||
| BC050398.2 | - | 4 - 5 | 311 | 76 | ||
| BC067364.1 | - | 3 - 4 | 296 | 6644672 | 76 | |
More Information
Set Membership: |
|

PRPF40B
Genomic Map