Assay Details
| Gene Symbol: SPG21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127889.4 | NP_001121361.1 | 2 - 3 | 190 | 115 | |
| NM_001127890.4 | NP_001121362.1 | 2 - 3 | 371 | 115 | ||
| NM_016630.6 | NP_057714.1 | 2 - 3 | 371 | 115 | ||
| XM_005254437.4 | XP_005254494.1 | 3 - 4 | 404 | 115 | ||
| XM_017022297.1 | XP_016877786.1 | 2 - 3 | 739 | 115 | ||
| XM_017022298.1 | XP_016877787.1 | 3 - 4 | 829 | 115 | ||
| GenBank mRNA | AF208861.1 | - | 1 - 2 | 86 | 115 | |
| AF212231.1 | - | 2 - 3 | 158 | 115 | ||
| AK172849.1 | - | 2 - 3 | 203 | 115 | ||
| AK225527.1 | - | 2 - 3 | 203 | 115 | ||
| AK301362.1 | - | 2 - 3 | 336 | 115 | ||
| AK312943.1 | - | 2 - 3 | 205 | 115 | ||
| BC000244.1 | - | 2 - 3 | 241 | 3357388 | 115 | |
| BI552731.1 | - | 2 - 3 | 341 | 115 | ||
| BI554234.1 | - | 3 - 4 | 399 | 115 | ||
| BI771887.1 | - | 2 - 3 | 205 | 115 | ||
| BP268459.1 | - | 2 - 3 | 364 | 115 | ||
| BP268875.1 | - | 2 - 3 | 364 | 115 | ||
| BX446863.2 | - | 2 - 3 | 321 | 115 | ||
| DA680695.1 | - | 2 - 3 | 189 | 115 | ||
More Information
Set Membership: |
|

SPG21
Genomic Map