Assay Details
| Gene Symbol: CCNO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021147.4 | NP_066970.3 | 1 - 2 | 579 | 112 | |
| GenBank mRNA | AK026075.1 | - | 1 - 2 | 542 | 112 | |
More Information
Set Membership: |
|

CCNO
Genomic Map