Assay Details
| Gene Symbol: TSEN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127394.3 | NP_001120866.1 | 2 - 3 | 324 | 94 | |
| NM_001300764.1 | NP_001287693.1 | 2 - 3 | 324 | 94 | ||
| NM_001300766.1 | NP_001287695.1 | 2 - 3 | 324 | 94 | ||
| NM_052965.3 | NP_443197.1 | 2 - 3 | 324 | 94 | ||
| NR_023349.2 | 2 - 3 | 324 | 94 | |||
| NR_125335.1 | 2 - 3 | 324 | 94 | |||
| XM_006711148.1 | XP_006711211.1 | 2 - 3 | 273 | 94 | ||
| XM_011509139.2 | XP_011507441.1 | 2 - 3 | 1768 | 94 | ||
| XM_017000228.1 | XP_016855717.1 | 2 - 3 | 1768 | 94 | ||
| XM_017000229.1 | XP_016855718.1 | 2 - 3 | 1768 | 94 | ||
| XM_017000230.1 | XP_016855719.1 | 2 - 3 | 1768 | 94 | ||
| GenBank mRNA | AF288394.1 | - | 2 - 3 | 299 | 94 | |
| AK098196.1 | - | 2 - 3 | 222 | 94 | ||
| AK296655.1 | - | 2 - 3 | 219 | 94 | ||
| BC022030.1 | - | 2 - 3 | 269 | 4824025 | 94 | |
| BI821111.1 | - | 2 - 3 | 249 | 94 | ||
| BM146150.1 | - | 2 - 3 | 326 | 94 | ||
| BM887891.1 | - | 2 - 3 | 156 | 94 | ||
| BU195337.1 | - | 2 - 3 | 296 | 94 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TSEN15
Genomic Map