Assay Details
| Gene Symbol: MED12L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053002.5 | NP_443728.3 | 37 - 38 | 5842 | 74 | |
| XM_006713487.3 | XP_006713550.1 | 39 - 40 | 6219 | 74 | ||
| XM_011512390.2 | XP_011510692.1 | 38 - 39 | 6114 | 74 | ||
| XM_017005681.1 | XP_016861170.1 | 24 - 25 | 3605 | 74 | ||
| GenBank mRNA | AB046855.1 | - | 24 - 25 | 3585 | 74 | |
| AF399708.1 | - | 37 - 38 | 5751 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000822.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000363124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000468305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000468305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000468305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000469768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000469768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000469768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000480026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000480026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000488092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000488092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |

MED12L
Genomic Map