Assay Details
| Gene Symbol: ZNF480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297625.1 | NP_001284554.1 | 3 - 4 | 276 | 108 | |
| NM_144684.3 | NP_653285.2 | 4 - 5 | 403 | 108 | ||
| XM_011526465.2 | XP_011524767.1 | 4 - 5 | 805 | 108 | ||
| GenBank mRNA | AK096559.1 | - | 4 - 5 | 393 | 108 | |
| AL832560.1 | - | 3 - 4 | 253 | 108 | ||
| AY512662.1 | - | 4 - 5 | 372 | 108 | ||
| BC007056.1 | - | 4 - 5 | 398 | 108 | ||
| BC018783.1 | - | 4 - 5 | 372 | 4873910 | 108 | |
| BC054018.1 | - | 4 - 5 | 403 | 108 | ||
| BC065503.1 | - | 4 - 5 | 378 | 108 | ||
| DA501214.1 | - | 4 - 5 | 401 | 108 | ||
More Information
Set Membership: |
|

ZNF480
Genomic Map