Assay Details
| Gene Symbol: SPG21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127889.4 | NP_001121361.1 | 4 - 5 | 430 | 146 | |
| NM_016630.6 | NP_057714.1 | 4 - 5 | 611 | 146 | ||
| XM_005254437.4 | XP_005254494.1 | 5 - 6 | 644 | 146 | ||
| XM_017022297.1 | XP_016877786.1 | 4 - 5 | 979 | 146 | ||
| XM_017022298.1 | XP_016877787.1 | 5 - 6 | 1069 | 146 | ||
| GenBank mRNA | AF208861.1 | - | 3 - 4 | 326 | 146 | |
| AF212231.1 | - | 4 - 5 | 398 | 146 | ||
| AK123725.1 | - | 2 - 3 | 843 | 146 | ||
| AK172849.1 | - | 4 - 5 | 443 | 146 | ||
| AK225527.1 | - | 4 - 5 | 443 | 146 | ||
| AK312943.1 | - | 4 - 5 | 445 | 146 | ||
| BC000244.1 | - | 4 - 5 | 481 | 3357388 | 146 | |
| BI554234.1 | - | 5 - 6 | 641 | 143 | ||
| BX446863.2 | - | 4 - 5 | 561 | 146 | ||
| DA680695.1 | - | 4 - 5 | 429 | 146 | ||
More Information
Set Membership: |
|

SPG21
Genomic Map