Assay Details
| Gene Symbol: PRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286356.1 | NP_001273285.1 | 1 - 2 | 379 | 108 | |
| GenBank mRNA | AK131573.1 | - | 1 - 2 | 379 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171884.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600171884.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171884.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600171884.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171884.hg.1 | TC1600009399.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171887.hg.1 | TC1600009399.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286357.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009399.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | ENST00000241808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | ENST00000435245.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | NM_001286356.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | NM_001286358.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | NM_001286359.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | NM_002762.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | NR_104428.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600171886.hg.1 | TC1600009399.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 144333 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 163342 | - | - |

PRM2
Genomic Map