Assay Details
| Gene Symbol: PRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286359.1 | NP_001273288.1 | 1 - 2 | 428 | 92 | |
| GenBank mRNA | DB066922.1 | - | 1 - 2 | 429 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

PRM2
Genomic Map