Assay Details
| Gene Symbol: PTTG1IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286822.1 | NP_001273751.1 | 1 - 2 | 401 | 62 | |
| NM_004339.3 | NP_004330.1 | 1 - 2 | 401 | 62 | ||
| NR_104597.1 | 1 - 2 | 401 | 62 | |||
| GenBank mRNA | AF149785.1 | - | 1 - 2 | 339 | 62 | |
| AK094882.1 | - | 1 - 2 | 134 | 62 | ||
| AK290139.1 | - | 1 - 2 | 154 | 62 | ||
| AK298557.1 | - | 1 - 2 | 189 | 62 | ||
| AK315626.1 | - | 1 - 2 | 216 | 62 | ||
| BC000415.2 | - | 1 - 2 | 158 | 2961420 | 62 | |
| BC012858.1 | - | 1 - 2 | 184 | 3857505 | 62 | |
| BC019295.1 | - | 1 - 2 | 148 | 2823814 | 62 | |
| BC020983.1 | - | 1 - 2 | 165 | 3851447 | 62 | |
| BC031097.1 | - | 1 - 2 | 179 | 5288388 | 62 | |
| BC034250.1 | - | 1 - 2 | 173 | 3862830 | 62 | |
| BG721942.1 | - | 1 - 2 | 340 | 62 | ||
| CN295711.1 | - | 1 - 2 | 192 | 62 | ||
| DC396020.1 | - | 1 - 2 | 402 | 62 | ||
| Z50022.1 | - | 1 - 2 | 207 | 62 | ||
More Information
Set Membership: |
|

PTTG1IP
Genomic Map