Assay Details
| Gene Symbol: GSTT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293814.1 | NP_001280743.1 | 1 - 2 | 192 | 63 | |
| GenBank mRNA | AJ867758.1 | - | 1 - 2 | 133 | 63 | |
More Information
Set Membership: |
|

GSTT1
Genomic Map