Assay Details
| Gene Symbol: PABPN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294328.1 | NP_001281257.1 | 4 - 5 | 620 | 78 | |
| GenBank mRNA | BC130001.1 | - | 4 - 5 | 620 | 40133435 | 78 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187546.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187546.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187546.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187546.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187546.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187546.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187546.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187546.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187546.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187546.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187546.hg.1 | TC1600011237.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |

PABPN1L
Genomic Map