Assay Details
| Gene Symbol: IL31RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242636.1 | NP_001229565.1 | 10 - 11 | 1537 | 79 | |
| NM_001242637.1 | NP_001229566.1 | 10 - 11 | 1546 | 79 | ||
| NM_001242638.1 | NP_001229567.1 | 12 - 13 | 1910 | 79 | ||
| NM_001242639.1 | NP_001229568.1 | 11 - 12 | 1681 | 79 | ||
| NM_001297570.1 | NP_001284499.1 | 10 - 11 | 1546 | 79 | ||
| NM_001297572.1 | NP_001284501.1 | 10 - 11 | 1674 | 79 | ||
| NM_139017.5 | NP_620586.3 | 10 - 11 | 1546 | 79 | ||
| XM_011543142.2 | XP_011541444.1 | 10 - 11 | 1379 | 79 | ||
| XM_011543143.2 | XP_011541445.1 | 10 - 11 | 1569 | 79 | ||
| XM_011543144.2 | XP_011541446.1 | 9 - 10 | 1451 | 79 | ||
| XM_011543145.2 | XP_011541447.1 | 7 - 8 | 1020 | 79 | ||
| GenBank mRNA | AF106913.1 | - | 10 - 11 | 1410 | 79 | |
| AF486620.1 | - | 9 - 10 | 1258 | 79 | ||
| AY358117.1 | - | 9 - 10 | 1258 | 79 | ||
| AY358740.1 | - | 10 - 11 | 1546 | 79 | ||
| AY499339.1 | - | 10 - 11 | 1419 | 79 | ||
| AY499341.1 | - | 10 - 11 | 1419 | 79 | ||
| AY499342.1 | - | 12 - 13 | 1793 | 79 | ||
| BC110490.2 | - | 11 - 12 | 1473 | 40007489 | 79 | |
More Information
Set Membership: |
|

IL31RA
Genomic Map