Assay Details
| Gene Symbol: MFAP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297710.1 | NP_001284639.1 | 2 - 3 | 326 | 95 | |
| GenBank mRNA | BX396041.2 | - | 2 - 3 | 125 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000359478.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000396549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000433590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000534833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000535336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000535411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000537009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000537128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000538107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000540087.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000543369.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000544211.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000544889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NM_001297709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NM_001297710.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NM_001297711.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NM_001297712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NM_003480.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NR_123733.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NR_123734.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000359478.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000396549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000433590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000534833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000535336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000535411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000537009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000537128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000538107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000540087.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000543369.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000544211.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | ENST00000544889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NM_001297709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NM_001297710.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NM_001297711.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NM_001297712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NM_003480.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NR_123733.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | NR_123734.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173961.hg.1 | TC1200009834.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000359478.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000396549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000433590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000534833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000535336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000535411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000537009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000537128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000538107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000538694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000540087.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000543369.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000544211.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000544889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NM_001297709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NM_001297710.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NM_001297711.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NM_001297712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NM_003480.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NR_123733.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NR_123734.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000359478.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000396549.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000433590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000534833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000535336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000535411.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000537009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000537128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000538107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000538694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000540087.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000543369.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000544211.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | ENST00000544889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NM_001297709.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NM_001297710.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NM_001297711.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NM_001297712.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NM_003480.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NR_123733.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | NR_123734.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200173958.hg.1 | TC1200009834.hg.1 | Clariom D TC |

MFAP5
Genomic Map