Assay Details
| Gene Symbol: NSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042549.1 | NP_001036014.1 | 1 - 2 | 272 | 75 | |
| NM_001297736.1 | NP_001284665.1 | 1 - 2 | 272 | 75 | ||
| NM_001297737.1 | NP_001284666.1 | 1 - 2 | 272 | 75 | ||
| NM_001297739.1 | NP_001284668.1 | 1 - 2 | 272 | 75 | ||
| NM_015471.3 | NP_056286.3 | 1 - 2 | 272 | 75 | ||
| GenBank mRNA | AF201941.1 | - | 1 - 2 | 244 | 75 | |
| AF255793.1 | - | 1 - 2 | 258 | 75 | ||
| AI703127.1 | - | 1 - 2 | 56 | 75 | ||
| AK222996.1 | - | 1 - 2 | 242 | 75 | ||
| AK223260.1 | - | 1 - 2 | 252 | 75 | ||
| AK303250.1 | - | 1 - 2 | 252 | 75 | ||
| AK313730.1 | - | 1 - 2 | 268 | 75 | ||
| AL050084.1 | - | 1 - 2 | 51 | 75 | ||
| BC007067.1 | - | 1 - 2 | 246 | 3997280 | 75 | |
| BC036048.1 | - | 1 - 2 | 250 | 75 | ||
| BE669670.1 | - | 1 - 2 | 77 | 75 | ||
| BU570983.1 | - | 1 - 2 | 252 | 75 | ||
| CK003923.1 | - | 1 - 2 | 255 | 75 | ||
| DB045932.1 | - | 1 - 2 | 272 | 75 | ||
More Information
Set Membership: |
|

NSL1
Genomic Map