Assay Details
| Gene Symbol: NSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297736.1 | NP_001284665.1 | 3 - 4 | 480 | 102 | |
| GenBank mRNA | AK303250.1 | - | 3 - 4 | 460 | 102 | |
| BE669670.1 | - | 3 - 4 | 285 | 102 | ||
More Information
Set Membership: |
|

NSL1
Genomic Map