Assay Details
| Gene Symbol: GPATCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297754.1 | NP_001284683.1 | 5 - 6 | 1230 | 144 | |
| GenBank mRNA | AK291238.1 | - | 5 - 6 | 1195 | 144 | |
| BC042193.1 | - | 5 - 6 | 1147 | 5532608 | 144 | |
| BC063474.1 | - | 5 - 6 | 1179 | 5206348 | 144 | |
More Information
Set Membership: |
|

GPATCH2
Genomic Map