Assay Details
| Gene Symbol: UGT2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105677.2 | NP_001099147.2 | 1 | 118 | 133 | |
| NM_001301233.1 | NP_001288162.1 | 1 | 118 | 133 | ||
| GenBank mRNA | FJ664272.1 | - | 1 | 118 | 133 | |
| FJ664273.1 | - | 1 | 118 | 133 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

UGT2A2
Genomic Map