Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006509.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006509.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006509.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006509.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006509.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006509.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006509.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006509.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006509.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006509.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006509.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400006509.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400144639.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400144639.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400144639.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400144639.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400144639.hg.1 |
TC0400006509.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400144641.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400144641.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400144641.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400144641.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400144641.hg.1 |
TC0400006509.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400144640.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400144640.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400144640.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400144640.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400144640.hg.1 |
TC0400006509.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400006509.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006509.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006509.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006509.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006509.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006509.hg.1 |
ENST00000625256.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400006509.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006509.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006509.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006509.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006509.hg.1 |
NR_104331.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400006509.hg.1 |
NR_104331.1 |
Refseq |