Assay Details
| Gene Symbol: SLC35F5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104470.1 | 15 - 16 | 2396 | 101 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RAB, member of RAS oncogene family-like 2A |
RABL2A | -71.8kb | Anti-sense | Chr. 2 - 113627218 - 113643398 on NCBI Build GRCh38 | |
| Downstream | microRNA 4782 | MIR4782 | 3.4kb | Sense | Chr. 2 - 113721290 - 113721368 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SLC35F5
Genomic Map