Assay Details
| Gene Symbol: DUSP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104474.1 | 1 - 2 | 510 | 75 | ||
| GenBank mRNA | DB177174.1 | - | 1 - 2 | 47 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC285766 | LOC285766 | -86.6kb | Anti-sense | Chr. 6 - 181466 - 205484 on NCBI Build GRCh38 | |
| Downstream | interferon regulatory factor 4 | IRF4 |
LSIRF, MUM1, NF-EM5, SHEP8 |
87.1kb | Sense | Chr. 6 - 391739 - 411443 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

DUSP22
Genomic Map