Assay Details
| Gene Symbol: ANKRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104587.1 | 6 - 7 | 1017 | 108 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 346 | LINC00346 |
C13orf29, NCRNA00346 |
-8.2kb | Sense | Chr. 13 - 110863987 - 110870308 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 431 | LINC00431 | 82.2kb | Anti-sense | Chr. 13 - 110966027 - 110990602 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

ANKRD10
Genomic Map