Assay Details
| Gene Symbol: CENPU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104593.1 | 10 - 11 | 998 | 124 | ||
| GenBank mRNA | BC067802.1 | - | 10 - 11 | 938 | 124 | |
| BC141854.1 | - | 10 - 11 | 996 | 8860370 | 124 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | primase and DNA directed polymerase | PRIMPOL |
CCDC111, MYP22 |
-2.7kb | Anti-sense | Chr. 4 - 184649613 - 184694959 on NCBI Build GRCh38 |
| Downstream | acyl-CoA synthetase long-chain family member 1 | ACSL1 |
ACS1, FACL1, FACL2, LACS, LACS1, LACS2 |
53.5kb | Sense | Chr. 4 - 184755595 - 184826593 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

CENPU
Genomic Map