| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015086.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015086.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015086.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015086.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212356.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212356.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212356.hg.1 |
TC0100015086.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100212357.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212357.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212357.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212357.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212357.hg.1 |
TC0100015086.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100015086.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015086.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015086.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015086.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015086.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212358.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212358.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212358.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212358.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212358.hg.1 |
TC0100015086.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100212360.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212360.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212360.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212360.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212360.hg.1 |
TC0100015086.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100212359.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212359.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212359.hg.1 |
ENST00000432195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100212359.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212359.hg.1 |
TC0100015086.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100212354.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212354.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212354.hg.1 |
TC0100015086.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100212355.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212355.hg.1 |
NR_104626.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100212355.hg.1 |
TC0100015086.hg.1 |
Clariom D TC |