Assay Details
| Gene Symbol: GLIPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104637.1 | 1 - 2 | 249 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | reversion inducing cysteine rich protein with kazal motifs | RECK | ST15 | -12.1kb | Sense | Chr. 9 - 36036905 - 36124455 on NCBI Build GRCh38 |
| Downstream | calicin | CCIN |
BTBD20, KBTBD14 |
27.3kb | Sense | Chr. 9 - 36169392 - 36171334 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

GLIPR2
Genomic Map