| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200197069.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200197069.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200197069.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200197069.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200197069.hg.1 |
TC1200012403.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200197070.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200197070.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200197070.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200197070.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200197070.hg.1 |
TC1200012403.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000536341.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
ENST00000540745.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200012403.hg.1 |
NR_104647.1 |
Refseq |