Assay Details
| Gene Symbol: LOC102467080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104659.1 | 1 - 2 | 159 | 91 | ||
| GenBank mRNA | AB371498.1 | - | 1 - 2 | 159 | 91 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BG621422.1 | - | 149 | Chr. - - [-] on Build | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105378967 | LOC105378967 | -18.0kb | Anti-sense | Chr. 5 - 54619445 - 54648062 on NCBI Build GRCh38 | |
| Downstream | endothelial cell specific molecule 1 | ESM1 | endocan | 274.5kb | Sense | Chr. 5 - 54977867 - 54985586 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC102467080
Genomic Map