Assay Details
| Gene Symbol: LOC102503427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105008.1 | 2 - 3 | 475 | 86 | ||
| GenBank mRNA | CA503367.1 | - | 2 - 3 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 366 | ZNF366 | DCSCRIPT | -62.4kb | Anti-sense | Chr. 5 - 72439899 - 72507422 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC102477328 | LOC102477328 | 2.6kb | Anti-sense | Chr. 5 - 72574075 - 72660669 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC102503427
Genomic Map