Assay Details
| Gene Symbol: LINC01333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105010.1 | 2 - 3 | 650 | 84 | ||
| GenBank mRNA | BC043537.1 | - | 2 - 3 | 650 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1335 | LINC01335 | -17.5kb | Anti-sense | Chr. 5 - 74306410 - 74308325 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1331 | LINC01331 | 41.0kb | Anti-sense | Chr. 5 - 74369376 - 74536976 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LINC01333
Genomic Map