| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011223.hg.1 |
ENST00000421004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011223.hg.1 |
ENST00000421004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011223.hg.1 |
ENST00000421004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011223.hg.1 |
ENST00000513755.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011223.hg.1 |
ENST00000513755.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011223.hg.1 |
ENST00000513755.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011223.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011223.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011223.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011223.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011223.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177890.hg.1 |
ENST00000421004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177890.hg.1 |
ENST00000513755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177890.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177890.hg.1 |
ENST00000421004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177890.hg.1 |
ENST00000513755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177890.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177890.hg.1 |
TC0500011223.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500177887.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177887.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177887.hg.1 |
TC0500011223.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500177889.hg.1 |
ENST00000421004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177889.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177889.hg.1 |
ENST00000421004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177889.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177889.hg.1 |
TC0500011223.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500177892.hg.1 |
ENST00000421004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177892.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177892.hg.1 |
ENST00000421004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500177892.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177892.hg.1 |
TC0500011223.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500177894.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177894.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500177894.hg.1 |
TC0500011223.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500011223.hg.1 |
ENST00000421004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011223.hg.1 |
ENST00000421004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011223.hg.1 |
ENST00000421004.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011223.hg.1 |
ENST00000513755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011223.hg.1 |
ENST00000513755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011223.hg.1 |
ENST00000513755.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011223.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011223.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011223.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011223.hg.1 |
NR_105014.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011223.hg.1 |
NR_105014.1 |
Refseq |