Assay Details
| Gene Symbol: LOC101927488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105040.1 | 1 - 2 | 187 | 181 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105369178 | LOC105369178 | Sense | Chr. 5 - 125980474 - 126368825 on NCBI Build GRCh38 | ||
| Downstream | GRAM domain containing 3 | GRAMD3 | NS3TP2 | 74.9kb | Anti-sense | Chr. 5 - 126360096 - 126494377 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101927488
Genomic Map