| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000553625.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000553625.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000553625.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008205.hg.1 |
NR_106822.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400158392.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400158392.hg.1 |
NR_106822.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400158392.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400158392.hg.1 |
NR_106822.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400158392.hg.1 |
TC1400008205.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000262238.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000553625.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000553625.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000553625.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000554804.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
ENST00000623799.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NM_003403.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008205.hg.1 |
NR_106822.1 |
Refseq |