Assay Details
| Gene Symbol: MIR6797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106855.1 | 1 | 35 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DMRT like family C2 | DMRTC2 | -17.3kb | Sense | Chr. 19 - 41844743 - 41852330 on NCBI Build GRCh38 | |
| Downstream | CD79a molecule | CD79A |
IGA, MB-1 |
7.4kb | Sense | Chr. 19 - 41877120 - 41881372 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1900008165.hg.1
Set Membership: |
|

MIR6797
Genomic Map