Assay Details
| Gene Symbol: MIR6830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106888.1 | 1 | 31 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | P4HA2 antisense RNA 1 | P4HA2-AS1 | -25.0kb | Anti-sense | Chr. 5 - 132184876 - 132192805 on NCBI Build GRCh38 | |
| Downstream | PDZ and LIM domain 4 | PDLIM4 | RIL | 39.7kb | Anti-sense | Chr. 5 - 132257658 - 132273454 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0500012006.hg.1
Set Membership: |
|

MIR6830
Genomic Map