Assay Details
| Gene Symbol: MIR7845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106999.1 | 1 | 53 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 2355 | MIR2355 | mir-2355 | -56.3kb | Anti-sense | Chr. 2 - 207109987 - 207110073 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927865 | LOC101927865 | 20.2kb | Anti-sense | Chr. 2 - 207186690 - 207254316 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0200010589.hg.1
Set Membership: |
|

MIR7845
Genomic Map