Assay Details
| Gene Symbol: MIR8056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_107023.1 | 1 | 44 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | stanniocalcin 2 | STC2 |
STC-2, STCRP |
-18.0kb | Anti-sense | Chr. 5 - 173314723 - 173329503 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC285593 | LOC285593 | 232.1kb | Sense | Chr. 5 - 173579634 - 173585068 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0500009499.hg.1
Set Membership: |
|

MIR8056
Genomic Map