Assay Details
| Gene Symbol: MIR8079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_107046.1 | 1 | 37 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small integral membrane protein 2 | SMIM2 | C13orf44 | -34.9kb | Sense | Chr. 13 - 44143151 - 44161257 on NCBI Build GRCh38 |
| Downstream | stress-associated endoplasmic reticulum protein family member 2 | SERP2 |
C13orf21, bA269C23.1 |
177.6kb | Anti-sense | Chr. 13 - 44373842 - 44397714 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC1300008782.hg.1
Set Membership: |
|

MIR8079
Genomic Map