| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006733.hg.1 |
ENST00000458168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006733.hg.1 |
ENST00000458168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006733.hg.1 |
NR_108039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006733.hg.1 |
NR_108039.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000006733.hg.1 |
NR_108039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000146125.hg.1 |
ENST00000458168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000146125.hg.1 |
NR_108039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000146125.hg.1 |
ENST00000458168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000146125.hg.1 |
NR_108039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000146125.hg.1 |
TC1000006733.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000146126.hg.1 |
ENST00000458168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000146126.hg.1 |
NR_108039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000146126.hg.1 |
ENST00000458168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000146126.hg.1 |
NR_108039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000146126.hg.1 |
TC1000006733.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000006733.hg.1 |
ENST00000458168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006733.hg.1 |
ENST00000458168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000006733.hg.1 |
NR_108039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006733.hg.1 |
NR_108039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000006733.hg.1 |
NR_108039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000146124.hg.1 |
NR_108039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000146124.hg.1 |
NR_108039.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000146124.hg.1 |
TC1000006733.hg.1 |
Clariom D TC |