Assay Details
| Gene Symbol: IL12A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108088.1 | 3 - 4 | 516 | 105 | ||
| GenBank mRNA | AK097161.1 | - | 3 - 4 | 516 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105374314 | LOC105374314 | Sense | Chr. 3 - 160032259 - 160040364 on NCBI Build GRCh38 | ||
| Downstream | chromosome 3 open reading frame 80 | C3orf80 | 36.4kb | Anti-sense | Chr. 3 - 160225636 - 160228213 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000462431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000462431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000462431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000462431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000462431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000486168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000486168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000486168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000486168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196832.hg.1 | ENST00000486168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196832.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196832.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196832.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196832.hg.1 | ENST00000486168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196832.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196832.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196832.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196832.hg.1 | TC0300012966.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300196828.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196828.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196828.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196828.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196828.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196828.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196828.hg.1 | TC0300012966.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300196830.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196830.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196830.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196830.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196830.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196830.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196830.hg.1 | TC0300012966.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300196833.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196833.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196833.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196833.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196833.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196833.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196833.hg.1 | TC0300012966.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000462431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000462431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000462431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000462431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000462431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000486168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000486168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000486168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000486168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012966.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196829.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196829.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196829.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196829.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196829.hg.1 | ENST00000497452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300196829.hg.1 | NR_108088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300196829.hg.1 | TC0300012966.hg.1 | Clariom D TC |

IL12A-AS1
Genomic Map