Assay Details
| Gene Symbol: IL12A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108088.1 | 8 - 9 | 1157 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | IQCJ-SCHIP1 readthrough | IQCJ-SCHIP1 | -91.7kb | Anti-sense | Chr. 3 - 159069252 - 159897366 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1100 | LINC01100 | 26.3kb | Anti-sense | Chr. 3 - 160020649 - 160031423 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

IL12A-AS1
Genomic Map