Assay Details
| Gene Symbol: LOC100422556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109775.1 | 2 - 3 | 277 | 94 | ||
| GenBank mRNA | HY269587.1 | - | 1 - 2 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tropomodulin 3 | TMOD3 | UTMOD | -6.8kb | Anti-sense | Chr. 15 - 51829628 - 51912138 on NCBI Build GRCh38 |
| Downstream |
LEO1 homolog, Paf1/RNA polymerase II complex component |
LEO1 | RDL | 15.1kb | Sense | Chr. 15 - 51938024 - 51971801 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100422556
Genomic Map