Assay Details
| Gene Symbol: LOC154449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002787.2 | 2 - 3 | 287 | 101 | ||
| GenBank mRNA | AK058183.1 | - | 2 - 3 | 287 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102724511 | LOC102724511 | -91.3kb | Anti-sense | Chr. 6 - 170160642 - 170163027 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC285804 | LOC285804 | 8.2kb | Sense | Chr. 6 - 170266669 - 170276762 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC154449
Genomic Map