Assay Details
| Gene Symbol: LOC101927100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_109875.1 | 2 - 3 | 408 | 99 | ||
| GenBank mRNA | DA313138.1 | - | 2 - 3 | 320 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TMED7-TICAM2 readthrough | TMED7-TICAM2 | Anti-sense | Chr. 5 - 115578496 - 115626179 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC102467217 | LOC102467217 | 21.7kb | Anti-sense | Chr. 5 - 115642125 - 115672652 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927100
Genomic Map