Assay Details
Gene Symbol: LOC101927379 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_109881.1 | 7 - 8 | 752 | 92 | ||
GenBank mRNA | BC043373.1 | - | 7 - 8 | 751 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101927357 | LOC101927357 | -112.6kb | Sense | Chr. 5 - 122581499 - 122584600 on NCBI Build GRCh38 | |
Downstream | sorting nexin 2 | SNX2 | TRG-9 | 75.1kb | Anti-sense | Chr. 5 - 122774996 - 122834539 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000508992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000508992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000508992.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000514657.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000514657.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000514657.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | ENST00000514657.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500181801.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500181801.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500181801.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500181801.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500181801.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500181801.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500181801.hg.1 | TC0500011864.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000508992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000508992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000508992.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000514657.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000514657.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000514657.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | ENST00000514657.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500011864.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500181802.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500181802.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500181802.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500181802.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500181802.hg.1 | ENST00000511194.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500181802.hg.1 | NR_109881.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500181802.hg.1 | TC0500011864.hg.1 | Clariom D TC |
